Search
Search

PACSIN1| Gene Symbol: PACSIN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001199583.2 | NP_001186512.1 | 5 | 819 | ||
| NM_020804.4 | NP_065855.1 | 5 | 832 | |||
| XM_011514541.1 | XP_011512843.1 | 5 | 687 | |||
| GenBank mRNA | AB037800.2 | - | 5 | 789 | ||
| AF242529.1 | - | 5 | 755 | |||
| AK289956.1 | - | 5 | 817 | |||
| AK294132.1 | - | 5 | 819 | |||
| AL834211.1 | - | 5 | 765 | |||
| BC040228.1 | - | 5 | 804 | 5173129 | ||
| BC128219.1 | - | 2 | 212 | |||