Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DEF6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022047.3 | NP_071330.3 | 2 | 253 | ||
GenBank mRNA | AJ276095.1 | - | 2 | 214 | ||
AK093613.1 | - | 2 | 253 | |||
AK314756.1 | - | 2 | 228 | |||
AY241694.1 | - | 2 | 253 | |||
BC017504.1 | - | 2 | 219 |