Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ARHGEF3 SIRNA Assays ›
Gene Symbol:
ARHGEF3
Gene Name:
Rho guanine nucleotide exchange factor 3
Gene Aliases:
GEF3, STA3, XPLN
Chromosome Location:
Chr.3: 56727418 - 57079308 on Build GRCh38
UniGene ID:
Hs.476402
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s27078
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
5,366.00
Your Price
Online offer:
5366.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ARHGEF3
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001128615.1
NP_001122087.1
10
1044
NM_001128616.1
NP_001122088.1
7
1460
NM_001289698.1
NP_001276627.1
8
968
NM_019555.2
NP_062455.1
7
1007
XM_005265186.4
XP_005265243.1
10
1108
XM_005265187.3
XP_005265244.1
10
1388
XM_011533764.1
XP_011532066.1
9
967
XM_011533765.1
XP_011532067.1
9
955
XM_011533766.2
XP_011532068.1
9
1299
XM_011533767.1
XP_011532069.1
8
919
XM_017006502.1
XP_016861991.1
12
1309
XM_017006503.1
XP_016861992.1
10
1044
XM_017006504.1
XP_016861993.1
10
1102
XM_017006505.1
XP_016861994.1
7
999
XM_017006506.1
XP_016861995.1
7
1261
GenBank mRNA
AB209661.1
-
7
1400
AF249744.1
-
7
965
AF433662.1
-
10
1001
AK024340.1
-
5
629
AK225688.1
-
7
989
AK291412.1
-
7
989
AK293864.1
-
7
1205
AK300045.1
-
8
968
AK302493.1
-
10
1044
AK308178.1
-
7
968
AL136832.1
-
7
942
AX775923.1
-
7
942
BC054345.1
-
7
955
6501674
BC068513.1
-
7
989
BC098122.1
-
7
960
40001657
BC098272.1
-
7
960
40001658
BC099715.1
-
6
883
40001659
BC104723.1
-
7
960
40001656
Back To Top
More Information
Panther Classification:
Molecular Function -
guanyl-nucleotide exchange factor
Gene Ontology Categories:
Function(s)
Process(es)
Rho protein signal transduction
regulation of Rho protein signal transduction
positive regulation of apoptotic process
positive regulation of GTPase activity
regulation of small GTPase mediated signal transduction
guanyl-nucleotide exchange factor activity
Rho guanyl-nucleotide exchange factor activity
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0