Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: HP1BP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016287.3 | NP_057371.2 | 12 | 1406 | ||
XM_005245875.4 | XP_005245932.1 | 12 | 1981 | |||
XM_005245876.3 | XP_005245933.1 | 12 | 1514 | |||
XM_005245877.4 | XP_005245934.1 | 12 | 1842 | |||
XM_005245878.4 | XP_005245935.1 | 12 | 1778 | |||
XM_005245879.4 | XP_005245936.1 | 11 | 1305 | |||
XM_011541532.1 | XP_011539834.1 | 12 | 1498 | |||
XM_011541533.1 | XP_011539835.1 | 12 | 1695 | |||
XM_011541534.2 | XP_011539836.1 | 11 | 1315 | |||
XM_017001393.1 | XP_016856882.1 | 12 | 1461 | |||
XM_017001394.1 | XP_016856883.1 | 12 | 1826 | |||
GenBank mRNA | AF113534.1 | - | 11 | 1339 | ||
AK023129.1 | - | 12 | 1406 | |||
AK291252.1 | - | 12 | 1384 | |||
AK303456.1 | - | 5 | 522 | |||
AK304065.1 | - | 11 | 1282 | |||
AL833978.1 | - | 8 | 1222 | |||
BC045660.1 | - | 8 | 2038 | 5271429 |