Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other NTM SIRNA Assays ›
          Gene Symbol:
          NTM
          Gene Name:
          neurotrimin
          Gene Aliases:
          HNT, IGLON2, NTRI
          Chromosome Location:
          Chr.11: 131370326 - 132336822 on Build GRCh38
          UniGene ID:
          Hs.504352
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s27176

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price 5,366.00
          Your Price
          Online offer:
          5366.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: NTM
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001048209.1 NP_001041674.1 6, 7 1110
          NM_001144058.1 NP_001137530.1 5, 6 1443
          NM_001144059.1 NP_001137531.1 5, 6 1443
          NM_016522.2 NP_057606.1 5, 6 1443
          XM_005271580.3 XP_005271637.1 6, 7 1260
          XM_005271581.3 XP_005271638.1 5, 6 1213
          XM_005271582.3 XP_005271639.1 6, 7 1260
          XM_006718855.3 XP_006718918.1 7, 8 1367
          XM_006718857.3 XP_006718920.1 6, 7 1287
          XM_011542861.2 XP_011541163.1 5, 6 1213
          XM_017017851.1 XP_016873340.1 5, 6 1213
          XM_017017852.1 XP_016873341.1 6, 7 1260
          XM_017017853.1 XP_016873342.1 7, 8 1367
          XM_017017854.1 XP_016873343.1 5, 6 1213
          XM_017017855.1 XP_016873344.1 6, 7 1260
          XM_017017856.1 XP_016873345.1 7, 8 973
          XM_017017857.1 XP_016873346.1 7, 8 1264
          XM_017017858.1 XP_016873347.1 7, 8 1257
          XM_017017859.1 XP_016873348.1 7, 8 973
          XM_017017860.1 XP_016873349.1 6, 7 1290
          XM_017017861.1 XP_016873350.1 7, 8 1367
          XM_017017862.1 XP_016873351.1 7, 8 1254
          XM_017017863.1 XP_016873352.1 6, 7 1290
          XM_017017864.1 XP_016873353.1 6, 7 1258
          XM_017017865.1 XP_016873354.1 6, 7 1260
          XM_017017866.1 XP_016873355.1 6, 7 1287
          GenBank mRNA AF126426.1 - 5, 6 1043
          AK293448.1 - 6, 7 1078
          AK293487.1 - 7, 8 1059
          AK294137.1 - 6, 7 989
          AK315964.1 - 6, 7 1105
          AY358331.1 - 6, 7 912
          BC050716.1 - 5, 6 793 6166839
          BE263639.1 - 4, 5 509
          EF043567.1 - 5, 6 799

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          immunoglobulin

          Gene Ontology Categories:

          Function(s) Process(es)

          cell adhesion
          neuron recognition
          protein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-5bf9q:80/100.66.78.48:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0