Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Instrument Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
Our Instagram
Our Facebook
Blog Behind the Bench
Customer Experience Center (CEC)
Ecommerce Exclusives
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other NTM SIRNA Assays ›
Gene Symbol:
NTM
Gene Name:
neurotrimin
Gene Aliases:
HNT, IGLON2, NTRI
Chromosome Location:
Chr.11: 131370326 - 132336822 on Build GRCh38
UniGene ID:
Hs.504352
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s27177
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
5,366.00
Your Price
Online offer:
5366.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: NTM
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001048209.1
NP_001041674.1
5
936
NM_001144058.1
NP_001137530.1
4
1269
NM_001144059.1
NP_001137531.1
4
1269
NM_016522.2
NP_057606.1
4
1269
XM_005271580.3
XP_005271637.1
5
1086
XM_005271581.3
XP_005271638.1
4
1039
XM_005271582.3
XP_005271639.1
5
1086
XM_006718855.3
XP_006718918.1
6
1193
XM_006718857.3
XP_006718920.1
5
1113
XM_011542861.2
XP_011541163.1
4
1039
XM_017017851.1
XP_016873340.1
4
1039
XM_017017852.1
XP_016873341.1
5
1086
XM_017017853.1
XP_016873342.1
6
1193
XM_017017854.1
XP_016873343.1
4
1039
XM_017017855.1
XP_016873344.1
5
1086
XM_017017856.1
XP_016873345.1
6
799
XM_017017857.1
XP_016873346.1
6
1090
XM_017017858.1
XP_016873347.1
6
1083
XM_017017859.1
XP_016873348.1
6
799
XM_017017860.1
XP_016873349.1
5
1116
XM_017017861.1
XP_016873350.1
6
1193
XM_017017862.1
XP_016873351.1
6
1080
XM_017017863.1
XP_016873352.1
5
1116
XM_017017864.1
XP_016873353.1
5
1084
XM_017017865.1
XP_016873354.1
5
1086
XM_017017866.1
XP_016873355.1
5
1113
XM_017017867.1
XP_016873356.1
3
293
GenBank mRNA
AF126426.1
-
4
869
AK293448.1
-
5
904
AK293487.1
-
6
885
AK294137.1
-
5
815
AK315964.1
-
5
931
AY358331.1
-
5
738
BC050716.1
-
4
619
6166839
BE263639.1
-
3
335
EF043567.1
-
4
625
Back To Top
More Information
Panther Classification:
Molecular Function -
immunoglobulin
Gene Ontology Categories:
Function(s)
Process(es)
cell adhesion
neuron recognition
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-5bf9q:80/100.66.78.48:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0