Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: SLC35B3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142541.2 | NP_001136013.1 | 9 | 1064 | ||
NM_015948.4 | NP_057032.2 | 9 | 1426 | |||
NR_109913.1 | 10 | 1417 | ||||
NR_109914.1 | 7 | 1360 | ||||
XM_005249156.3 | XP_005249213.1 | 8 | 1206 | |||
XM_005249157.2 | XP_005249214.1 | 9 | 1199 | |||
XM_005249158.1 | XP_005249215.1 | 8 | 1094 | |||
XM_006715102.3 | XP_006715165.1 | 9 | 1236 | |||
XM_006715103.3 | XP_006715166.1 | 7 | 1113 | |||
XM_011514648.1 | XP_011512950.1 | 10 | 1206 | |||
XM_017010910.1 | XP_016866399.1 | 9 | 1522 | |||
XM_017010911.1 | XP_016866400.1 | 8 | 1440 | |||
XM_017010912.1 | XP_016866401.1 | 8 | 1061 | |||
XM_017010913.1 | XP_016866402.1 | 8 | 1429 | |||
XM_017010914.1 | XP_016866403.1 | 8 | 1290 | |||
XM_017010915.1 | XP_016866404.1 | 7 | 1001 | |||
GenBank mRNA | AB231931.1 | - | 7 | 901 | ||
AF132953.1 | - | 8 | 922 | |||
AK313522.1 | - | 9 | 1042 | |||
BC006973.1 | - | 9 | 1252 | 3827889 | ||
BX538271.1 | - | 10 | 1412 | |||
BX641086.1 | - | 8 | 6565 | |||
BX647208.1 | - | 7 | 1294 | |||
BX648022.1 | - | 8 | 5363 |