Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: HDDC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016063.2 | NP_057147.2 | 5 | 633 | ||
GenBank mRNA | AF151888.1 | - | 4 | 390 | ||
AF529363.1 | - | 5 | 429 | |||
BC001671.2 | - | 4 | 465 | |||
BC003357.1 | - | 5 | 472 | |||
BC066332.1 | - | 6 | 447 | 5294646 | ||
BE743302.1 | - | 5 | 546 | |||
BP257644.1 | - | 1 | 20 |