Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: GLRX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243399.1 | NP_001230328.1 | 2 | 452 | ||
NM_001319291.1 | NP_001306220.1 | 2 | 33 | |||
NM_016066.4 | NP_057150.2 | 2 | 610 | |||
NM_197962.2 | NP_932066.1 | 2 | 227 | |||
XM_017001397.1 | XP_016856886.1 | 2 | 160 | |||
GenBank mRNA | AF132495.2 | - | 2 | 142 | ||
AF151891.1 | - | 2 | 117 | |||
AF290514.1 | - | 2 | 610 | |||
AY038988.1 | - | 2 | 149 | |||
BC009669.1 | - | 2 | 142 | |||
BC028113.1 | - | 2 | 133 | 5395272 | ||
BM887926.1 | - | 2 | 33 | |||
BQ432881.1 | - | 2 | 458 | |||
BU157087.1 | - | 182 | ||||
CN362908.1 | - | 2 | 251 |