Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: MEMO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001137602.2 | NP_001131074.1 | 6, 7 | 1013 | ||
NM_001301833.1 | NP_001288762.1 | 8, 9 | 793 | |||
NM_001301852.1 | NP_001288781.1 | 6, 7 | 572 | |||
NM_015955.3 | NP_057039.1 | 7, 8 | 1082 | |||
XM_011532892.2 | XP_011531194.1 | 8, 9 | 730 | |||
XM_011532896.2 | XP_011531198.1 | 5, 6 | 465 | |||
XM_017004255.1 | XP_016859744.1 | 8, 9 | 703 | |||
GenBank mRNA | AB041018.1 | - | 7, 8 | 1082 | ||
AB041019.1 | - | 7, 8 | 731 | |||
AF132961.1 | - | 7, 8 | 780 | |||
AF363446.1 | - | 7, 8 | 999 | |||
AF529368.1 | - | 7, 8 | 665 | |||
AK290753.1 | - | 8, 9 | 749 | |||
AK297128.1 | - | 6, 7 | 724 | |||
BC018733.1 | - | 7, 8 | 1047 | 4851517 | ||
BC070046.1 | - | 8, 9 | 822 | 5273070 | ||
BC094681.1 | - | 7, 8 | 686 | 6205399 | ||
BG033287.1 | - | 6, 7 | 520 |