Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • Enterprise Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other CDC20 SIRNA Assays ›
          Gene Symbol:
          CDC20
          Gene Name:
          cell division cycle 20
          Gene Aliases:
          CDC20A, bA276H19.3, p55CDC
          Chromosome Location:
          Chr.1: 43358955 - 43363203 on Build GRCh38
          UniGene ID:
          Hs.524947
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s2747

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: CDC20
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001255.2 NP_001246.2 8 982
          GenBank mRNA AF099644.1 - 7 925
          AK225504.1 - 8 961
          AK303844.1 - 9 937
          AK312780.1 - 8 961
          BC000624.2 - 7 989 3346682
          BC001088.2 - 8 982 2964686
          BC006272.2 - 7 924 3948819
          BC009425.1 - 8 964 2959460
          BC009426.1 - 8 964 2959596
          BC010044.2 - 8 973 3634656
          BC012803.2 - 8 964 2959588
          BC012827.2 - 8 982 2964686
          BC013303.2 - 8 964 2959588
          BC015998.1 - 8 960 4306426
          BC024257.1 - 8 964 2959460
          BC031294.1 - 8 964 2959487
          BC110321.1 - 7 945 5496957
          BT007388.1 - 7 881
          U05340.1 - 8 991

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Mouse: NM_023223, AK188208, BC003215, AK160204, AK029424, and others.

          Panther Classification:

          Molecular Function -

          ubiquitin-protein ligase

          Gene Ontology Categories:

          Function(s) Process(es)

          cell cycle
          sister chromatid cohesion
          mitotic nuclear division
          positive regulation of cell proliferation
          anaphase-promoting complex-dependent catabolic process
          positive regulation of synaptic plasticity
          regulation of meiotic nuclear division
          protein ubiquitination involved in ubiquitin-dependent protein catabolic process
          proteasome-mediated ubiquitin-dependent protein catabolic process
          regulation of dendrite development
          cell division
          negative regulation of ubiquitin-protein ligase activity involved in mitotic cell cycle
          positive regulation of ubiquitin-protein ligase activity involved in regulation of mitotic cell cycle transition
          regulation of ubiquitin-protein ligase activity involved in mitotic cell cycle
          positive regulation of synapse maturation
          positive regulation of ubiquitin protein ligase activity
          protein binding
          protein C-terminus binding
          anaphase-promoting complex binding
          enzyme binding
          histone deacetylase binding
          ubiquitin-protein transferase activator activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0