Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: SS18L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016305.3 | NP_057389.1 | 4 | 826 | ||
XM_017006548.1 | XP_016862037.1 | 3 | 1838 | |||
XM_017006549.1 | XP_016862038.1 | 4 | 797 | |||
GenBank mRNA | AF201950.1 | - | 3 | 716 | ||
BC017804.1 | - | 3 | 644 | 4658643 | ||
BC020444.1 | - | 3 | 616 |