Search
Search

COQ3| Gene Symbol: COQ3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017421.3 | NP_059117.3 | 4, 5 | 509 | ||
| XM_006715500.3 | XP_006715563.1 | 5, 6 | 648 | |||
| XM_011535899.1 | XP_011534201.1 | 3, 4 | 337 | |||
| XM_017010933.1 | XP_016866422.1 | 5, 6 | 506 | |||
| GenBank mRNA | AF193016.1 | - | 4, 5 | 446 | ||
| AF351615.1 | - | 4, 5 | 514 | |||
| AK056955.1 | - | 4, 5 | 507 | |||
| AL136726.1 | - | 4, 5 | 507 | |||
| BC001534.1 | - | 4, 5 | 480 | |||
| BC015634.2 | - | 4, 5 | 470 | |||
| BC063463.1 | - | 4, 5 | 485 | 5196692 | ||