Search
Search

COQ3| Gene Symbol: COQ3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017421.3 | NP_059117.3 | 5 | 566 | ||
| XM_006715500.3 | XP_006715563.1 | 6 | 705 | |||
| XM_011535899.1 | XP_011534201.1 | 4 | 394 | |||
| XM_017010933.1 | XP_016866422.1 | 6 | 563 | |||
| GenBank mRNA | AF193016.1 | - | 5 | 503 | ||
| AF351615.1 | - | 5 | 571 | |||
| AK056955.1 | - | 5 | 564 | |||
| BC001534.1 | - | 5 | 537 | |||
| BC015634.2 | - | 5 | 527 | |||
| BC063463.1 | - | 5 | 542 | 5196692 | ||