Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CCHCR1 SIRNA Assays ›
Gene Symbol:
CCHCR1
Gene Name:
coiled-coil alpha-helical rod protein 1
Gene Aliases:
C6orf18, HCR, SBP
Chromosome Location:
Chr.6: 31142439 - 31158238 on Build GRCh38
UniGene ID:
Hs.485075
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s29151
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CCHCR1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001105563.1
NP_001099033.1
17
2477
NM_001105564.1
NP_001099034.1
17
2585
NM_019052.3
NP_061925.2
17
2252
XM_011514702.2
XP_011513004.1
17
2511
XM_011514703.1
XP_011513005.1
18
2324
XM_011514704.2
XP_011513006.1
17
2113
XM_011514705.1
XP_011513007.1
17
2222
XM_017010961.1
XP_016866450.1
17
2619
XM_017010962.1
XP_016866451.1
18
2371
XM_017010963.1
XP_016866452.1
18
2485
XM_017010964.1
XP_016866453.1
17
2249
XM_017010965.1
XP_016866454.1
17
2311
XM_017010966.1
XP_016866455.1
17
2332
XM_017010967.1
XP_016866456.1
17
2435
XM_017010968.1
XP_016866457.1
17
2204
XM_017010969.1
XP_016866458.1
17
2111
XM_017010970.1
XP_016866459.1
16
1897
GenBank mRNA
AB029331.1
-
15
2073
AB112474.1
-
17
2208
AB112475.1
-
17
2559
AF216493.1
-
15
2073
AK000204.1
-
17
2208
AK000217.1
-
17
2209
AK000533.1
-
12
1324
AK289446.1
-
17
2200
AK295494.1
-
17
2230
AY029160.1
-
16
2129
BC110534.1
-
17
2295
40035162
BC110535.1
-
17
2408
40035164
CR457227.1
-
16
2129
Back To Top
More Information
Gene Ontology Categories:
Function(s)
protein export from nucleus
multicellular organism development
cell differentiation
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-7szzr:80/100.66.78.37:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0