Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other RBFOX1 SIRNA Assays ›
          Gene Symbol:
          RBFOX1
          Gene Name:
          RNA binding protein, fox-1 homolog 1
          Gene Aliases:
          2BP1, A2BP1, FOX-1, FOX1, HRNBP1
          Chromosome Location:
          Chr.16: 5239749 - 7713343 on Build GRCh38
          UniGene ID:
          Hs.459842
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s29338

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: RBFOX1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001142333.1 NP_001135805.1 6, 7 1397
          NM_001142334.1 NP_001135806.1 4, 5 629
          NM_001308117.1 NP_001295046.1 6, 7 734
          NM_018723.3 NP_061193.2 6, 7 1397
          NM_145891.2 NP_665898.1 3, 4 721
          NM_145892.2 NP_665899.1 3, 4 721
          NM_145893.2 NP_665900.1 3, 4 721
          XM_005255386.3 XP_005255443.1 3, 4 647
          XM_005255387.3 XP_005255444.1 3, 4 626
          XM_005255388.4 XP_005255445.1 3, 4 682
          XM_005255390.3 XP_005255447.1 6, 7 1519
          XM_005255391.3 XP_005255448.1 5, 6 529
          XM_005255394.4 XP_005255451.1 3, 4 626
          XM_011522546.2 XP_011520848.1 3, 4 626
          XM_011522547.2 XP_011520849.1 3, 4 626
          XM_011522548.2 XP_011520850.1 3, 4 626
          XM_017023318.1 XP_016878807.1 7, 8 1024
          XM_017023319.1 XP_016878808.1 9, 10 1020
          XM_017023320.1 XP_016878809.1 6, 7 1428
          XM_017023321.1 XP_016878810.1 5, 6 2490
          XM_017023322.1 XP_016878811.1 5, 6 2490
          XM_017023323.1 XP_016878812.1 3, 4 647
          XM_017023324.1 XP_016878813.1 6, 7 1428
          XM_017023325.1 XP_016878814.1 4, 5 1026
          XM_017023326.1 XP_016878815.1 6, 7 1519
          XM_017023327.1 XP_016878816.1 3, 4 647
          XM_017023328.1 XP_016878817.1 6, 7 1428
          XM_017023329.1 XP_016878818.1 6, 7 1428
          XM_017023330.1 XP_016878819.1 5, 6 1967
          XM_017023331.1 XP_016878820.1 3, 4 2433
          XM_017023332.1 XP_016878821.1 3, 4 647
          XM_017023333.1 XP_016878822.1 5, 6 529
          XM_017023334.1 XP_016878823.1 3, 4 647
          XM_017023335.1 XP_016878824.1 6, 7 1519
          XM_017023336.1 XP_016878825.1 5, 6 529
          XM_017023337.1 XP_016878826.1 3, 4 647
          XM_017023338.1 XP_016878827.1 5, 6 529
          XM_017023339.1 XP_016878828.1 3, 4 673
          XM_017023340.1 XP_016878829.1 5, 6 529
          XM_017023341.1 XP_016878830.1 6, 7 1519
          XM_017023342.1 XP_016878831.1 5, 6 529
          XM_017023343.1 XP_016878832.1 3, 4 676
          GenBank mRNA AB208826.1 - 1431
          AF094849.1 - 3, 4 722
          AF107203.1 - 6, 7 1395
          AF109106.1 - 3, 4 722
          AF229057.1 - 3, 4 724
          AF448859.1 - 6, 7 1395
          AK001027.1 - 4, 5 629
          AK293951.1 - 6, 7 734
          AL713700.1 - 5, 6 1244
          BC113691.1 - 3, 4 515 8322743
          BC143816.1 - 3, 4 515
          KJ534926.1 - 3, 4 469
          KJ535000.1 - 3, 4 469
          KJ535001.1 - 3, 4 469

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          RNA metabolism protein RNA processing factor RNA splicing factor nucleic acid metabolism protein

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of alternative mRNA splicing, via spliceosome
          mRNA processing
          nervous system development
          RNA splicing
          RNA transport
          nucleotide binding
          RNA binding
          mRNA binding
          protein binding
          protein C-terminus binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-7ngt7:80/100.66.72.242:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0