Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
CDMO and Clinical Trial Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other BCAS3 SIRNA Assays ›
Gene Symbol:
BCAS3
Gene Name:
BCAS3, microtubule associated cell migration factor
Gene Aliases:
GAOB1, MAAB
Chromosome Location:
Chr.17: 60677811 - 61392838 on Build GRCh38
UniGene ID:
Hs.655028
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s29532
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: BCAS3
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001099432.1
NP_001092902.1
12
1074
NM_001320470.1
NP_001307399.1
12
1074
NM_017679.3
NP_060149.3
12
1074
XM_005257472.1
XP_005257529.1
12
1025
XM_011524939.1
XP_011523241.1
12
1025
XM_011524940.1
XP_011523242.1
12
1025
XM_011524941.1
XP_011523243.1
12
1025
XM_011524942.2
XP_011523244.1
12
1025
XM_011524943.2
XP_011523245.1
12
1025
XM_011524944.1
XP_011523246.1
12
1025
XM_011524945.2
XP_011523247.1
12
1025
XM_011524946.2
XP_011523248.1
12
1025
XM_011524947.1
XP_011523249.1
12
1025
XM_011524948.2
XP_011523250.1
12
1025
XM_011524949.2
XP_011523251.1
12
1025
XM_011524950.1
XP_011523252.1
12
1025
XM_011524951.2
XP_011523253.1
12
1025
XM_017024783.1
XP_016880272.1
12
1025
XM_017024784.1
XP_016880273.1
12
1025
XM_017024785.1
XP_016880274.1
12
1025
XM_017024786.1
XP_016880275.1
12
1025
XM_017024787.1
XP_016880276.1
12
1025
XM_017024788.1
XP_016880277.1
12
1025
XM_017024789.1
XP_016880278.1
12
1025
XM_017024790.1
XP_016880279.1
12
1025
XM_017024791.1
XP_016880280.1
12
1025
XM_017024792.1
XP_016880281.1
12
1025
XM_017024793.1
XP_016880282.1
12
1025
XM_017024794.1
XP_016880283.1
12
1025
XM_017024795.1
XP_016880284.1
12
1025
XM_017024796.1
XP_016880285.1
4
451
GenBank mRNA
AF361219.1
-
13
1013
AJ518105.1
-
4
466
AJ518836.1
-
4
381
AK022526.1
-
4
429
AK023054.1
-
4
421
AK025510.1
-
12
990
AK225757.1
-
12
1033
AK304791.1
-
5
433
AK314744.1
-
12
1019
BC017390.1
-
12
1030
BC117275.1
-
12
996
40125826
BC143386.1
-
12
996
9051892
BC143387.1
-
12
996
9051893
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
angiogenesis
transcription, DNA-templated
Golgi organization
positive regulation of endothelial cell migration
vesicle-mediated transport
microtubule organizing center organization
negative regulation of GTPase activity
tube formation
response to starvation
positive regulation of catalytic activity
positive regulation of GTPase activity
positive regulation of transcription from RNA polymerase II promoter
positive regulation of filopodium assembly
negative regulation of focal adhesion assembly
cellular response to estrogen stimulus
positive regulation of intracellular protein transport
activation of GTPase activity
regulation of establishment of cell polarity
positive regulation of actin cytoskeleton reorganization
chromatin binding
protein binding
transcription factor binding
acetyltransferase activator activity
histone acetyltransferase binding
nuclear hormone receptor binding
histone binding
beta-tubulin binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-74fd55b88b-zl9nd:80/10.239.167.245:80.
git-commit: e4d250c089da5f210ba2ef291b70bc15861d50fd
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.9.0-2024.05.21-1.0