Search
Search

BSPRY| Gene Symbol: BSPRY | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001317943.1 | NP_001304872.1 | 5 | 666 | ||
| NM_017688.2 | NP_060158.2 | 5 | 651 | |||
| XM_006717149.3 | XP_006717212.1 | 5 | 639 | |||
| GenBank mRNA | AJ276691.1 | - | 5 | 632 | ||
| AK000157.1 | - | 5 | 377 | |||
| AK092607.1 | - | 5 | 642 | |||
| BP370567.1 | - | 5 | 489 | |||
| BX462607.2 | - | 5 | 650 | |||