Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: CC2D1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017721.4 | NP_060191.3 | 21 | 2496 | ||
XM_005259972.2 | XP_005260029.1 | 21 | 2441 | |||
XM_005259973.2 | XP_005260030.1 | 21 | 2444 | |||
XM_005259974.2 | XP_005260031.1 | 21 | 2432 | |||
XM_005259975.2 | XP_005260032.1 | 21 | 2429 | |||
GenBank mRNA | AB097002.1 | - | 21 | 2438 | ||
AF536205.1 | - | 23 | 2549 | |||
AK023399.1 | - | 14 | 1321 | |||
AK026371.1 | - | 5 | 334 | |||
AK315068.1 | - | 21 | 2414 | |||
BC006556.2 | - | 16 | 1600 | |||
BC048345.1 | - | 7 | 508 | |||
BC064981.1 | - | 21 | 2400 | 6585236 |