Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: PALMD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017734.4 | NP_060204.1 | 3 | 656 | ||
GenBank mRNA | AF262379.1 | - | 3 | 587 | ||
AJ312214.1 | - | 3 | 498 | |||
AK000278.1 | - | 3 | 418 | |||
AL390971.1 | - | 3 | 586 | |||
AL390973.1 | - | 3 | 382 | |||
BC012570.1 | - | 3 | 471 | 4245261 | ||
KU178657.1 | - | 3 | 213 | |||
KU178658.1 | - | 3 | 213 |