Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other PXK SIRNA Assays ›
          Gene Symbol:
          PXK
          Gene Name:
          PX domain containing serine/threonine kinase like
          Gene Aliases:
          MONaKA
          Chromosome Location:
          Chr.3: 58332868 - 58426127 on Build GRCh38
          UniGene ID:
          Hs.190544
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s29711
          Validated

          Purification
          Size
          Availability Inventoried
          Catalog # 4390824
          Price 7,898.00
          Your Price
          Online offer:
          7898.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: PXK
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001289095.1 NP_001276024.1 7, 8 761
          NM_001289096.1 NP_001276025.1 6, 7 713
          NM_001289098.1 NP_001276027.1 8, 9 812
          NM_001289099.1 NP_001276028.1 8, 9 917
          NM_001289100.1 NP_001276029.1 6, 7 713
          NM_001289101.1 NP_001276030.1 5, 6 526
          NM_017771.4 NP_060241.2 8, 9 812
          XM_005265250.4 XP_005265307.1 8, 9 812
          XM_005265252.4 XP_005265309.1 8, 9 812
          XM_005265255.3 XP_005265312.1 8, 9 812
          XM_005265256.3 XP_005265313.1 8, 9 812
          XM_017006671.1 XP_016862160.1 7, 8 703
          XM_017006672.1 XP_016862161.1 7, 8 703
          XM_017006673.1 XP_016862162.1 6, 7 704
          XM_017006674.1 XP_016862163.1 7, 8 703
          XM_017006675.1 XP_016862164.1 8, 9 812
          XM_017006676.1 XP_016862165.1 8, 9 812
          XM_017006677.1 XP_016862166.1 8, 9 812
          XM_017006678.1 XP_016862167.1 8, 9 812
          XM_017006679.1 XP_016862168.1 8, 9 812
          XM_017006680.1 XP_016862169.1 8, 9 812
          XM_017006681.1 XP_016862170.1 7, 8 703
          XM_017006682.1 XP_016862171.1 7, 8 703
          XM_017006683.1 XP_016862172.1 7, 8 703
          XM_017006684.1 XP_016862173.1 7, 8 703
          XM_017006685.1 XP_016862174.1 7, 8 703
          XM_017006686.1 XP_016862175.1 9, 10 926
          XM_017006687.1 XP_016862176.1 9, 10 924
          XM_017006688.1 XP_016862177.1 6, 7 688
          XM_017006689.1 XP_016862178.1 6, 7 678
          XM_017006690.1 XP_016862179.1 6, 7 669
          XM_017006691.1 XP_016862180.1 5, 6 525
          XM_017006692.1 XP_016862181.1 9, 10 963
          XM_017006693.1 XP_016862182.1 9, 10 963
          XM_017006694.1 XP_016862183.1 9, 10 857
          XM_017006695.1 XP_016862184.1 9, 10 963
          XM_017006696.1 XP_016862185.1 5, 6 525
          XM_017006697.1 XP_016862186.1 5, 6 520
          XM_017006698.1 XP_016862187.1 5, 6 518
          XM_017006699.1 XP_016862188.1 5, 6 530
          XM_017006700.1 XP_016862189.1 5, 6 548
          XM_017006701.1 XP_016862190.1 5, 6 525
          XM_017006702.1 XP_016862191.1 5, 6 525
          XM_017006703.1 XP_016862192.1 5, 6 525
          XM_017006704.1 XP_016862193.1 5, 6 525
          GenBank mRNA AB209688.1 - 8, 9 790
          AF399753.1 - 8, 9 713
          AK000342.1 - 8, 9 810
          AK131385.1 - 7, 8 703
          AK295542.1 - 6, 7 705
          AK296034.1 - 5, 6 384
          AK296215.1 - 5, 6 461
          AY274811.1 - 8, 9 812
          AY437879.1 - 6, 7 656
          AY847220.1 - 8, 9 820
          AY847221.1 - 5, 6 429
          AY847222.1 - 5, 6 488
          BC008943.2 - 8, 9 793
          BC014479.1 - 8, 9 728 4856139
          DQ124707.1 - 8, 9 713
          DQ124708.1 - 8, 9 713
          KF774202.1 - 8, 9 822
          KF774203.1 - 8, 9 822
          KF774204.1 - 8, 9 897

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_182821, XM_008770535, XM_008770538, XM_008770532, XM_006251775, and others.
          This siRNA also targets the following transcripts in Mouse: NM_145458, NM_178279, AK152789, AK172347, AK166036, and others.

          Matching Taqman Gene Expression Assay:


          Hs00298958_m1

          Gene Ontology Categories:

          Function(s) Process(es)

          protein phosphorylation
          inflammatory response
          negative regulation of ATPase activity
          regulation of membrane potential
          negative regulation of ion transport
          modulation of synaptic transmission
          nucleotide binding
          actin binding
          protein kinase activity
          ATP binding
          protein C-terminus binding
          phosphatidylinositol binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-8bdhq:80/100.66.78.62:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0