Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PXK SIRNA Assays ›
Gene Symbol:
PXK
Gene Name:
PX domain containing serine/threonine kinase like
Gene Aliases:
MONaKA
Chromosome Location:
Chr.3: 58332868 - 58426127 on Build GRCh38
UniGene ID:
Hs.190544
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s29712
Validated
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
1 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
1 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
1 umol
10 umol
Availability
Inventoried
Catalog #
4390824
Price
7,898.00
Your Price
Online offer:
7898.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PXK
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001289095.1
NP_001276024.1
4, 5
516
NM_001289096.1
NP_001276025.1
3, 4
468
NM_001289098.1
NP_001276027.1
5, 6
567
NM_001289099.1
NP_001276028.1
5, 6
672
NM_001289100.1
NP_001276029.1
3, 4
468
NM_001289101.1
NP_001276030.1
2, 3
281
NM_017771.4
NP_060241.2
5, 6
567
XM_005265250.4
XP_005265307.1
5, 6
567
XM_005265252.4
XP_005265309.1
5, 6
567
XM_005265255.3
XP_005265312.1
5, 6
567
XM_005265256.3
XP_005265313.1
5, 6
567
XM_017006671.1
XP_016862160.1
4, 5
458
XM_017006672.1
XP_016862161.1
4, 5
458
XM_017006673.1
XP_016862162.1
3, 4
459
XM_017006674.1
XP_016862163.1
4, 5
458
XM_017006675.1
XP_016862164.1
5, 6
567
XM_017006676.1
XP_016862165.1
5, 6
567
XM_017006677.1
XP_016862166.1
5, 6
567
XM_017006678.1
XP_016862167.1
5, 6
567
XM_017006679.1
XP_016862168.1
5, 6
567
XM_017006680.1
XP_016862169.1
5, 6
567
XM_017006681.1
XP_016862170.1
4, 5
458
XM_017006682.1
XP_016862171.1
4, 5
458
XM_017006683.1
XP_016862172.1
4, 5
458
XM_017006684.1
XP_016862173.1
4, 5
458
XM_017006685.1
XP_016862174.1
4, 5
458
XM_017006686.1
XP_016862175.1
6, 7
681
XM_017006687.1
XP_016862176.1
6, 7
679
XM_017006688.1
XP_016862177.1
3, 4
443
XM_017006689.1
XP_016862178.1
3, 4
433
XM_017006690.1
XP_016862179.1
3, 4
424
XM_017006691.1
XP_016862180.1
2, 3
280
XM_017006692.1
XP_016862181.1
6, 7
718
XM_017006693.1
XP_016862182.1
6, 7
718
XM_017006694.1
XP_016862183.1
6, 7
612
XM_017006695.1
XP_016862184.1
6, 7
718
XM_017006696.1
XP_016862185.1
2, 3
280
XM_017006697.1
XP_016862186.1
2, 3
275
XM_017006698.1
XP_016862187.1
2, 3
273
XM_017006699.1
XP_016862188.1
2, 3
285
XM_017006700.1
XP_016862189.1
2, 3
303
XM_017006701.1
XP_016862190.1
2, 3
280
XM_017006702.1
XP_016862191.1
2, 3
280
XM_017006703.1
XP_016862192.1
2, 3
280
XM_017006704.1
XP_016862193.1
2, 3
280
GenBank mRNA
AB209688.1
-
5, 6
545
AF399753.1
-
5, 6
468
AK000342.1
-
5, 6
565
AK131385.1
-
4, 5
458
AK295542.1
-
3, 4
460
AK296034.1
-
2, 3
139
AK296215.1
-
2, 3
216
AY274811.1
-
5, 6
567
AY437879.1
-
3, 4
411
AY847220.1
-
5, 6
575
AY847221.1
-
2, 3
184
BC008943.2
-
5, 6
548
BC014479.1
-
5, 6
483
4856139
DQ124707.1
-
5, 6
468
DQ124708.1
-
5, 6
468
KF774202.1
-
5, 6
577
KF774203.1
-
5, 6
577
KF774204.1
-
5, 6
652
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_182821, XM_008770535, XM_008770538, XM_008770532, XM_006251775, and others.
Matching Taqman Gene Expression Assay:
Hs00298958_m1
Gene Ontology Categories:
Function(s)
Process(es)
protein phosphorylation
inflammatory response
negative regulation of ATPase activity
regulation of membrane potential
negative regulation of ion transport
modulation of synaptic transmission
nucleotide binding
actin binding
protein kinase activity
ATP binding
protein C-terminus binding
phosphatidylinositol binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-4b6rj:80/100.66.75.107:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0