Search Thermo Fisher Scientific
Gene Symbol: ATG16L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190266.1 | NP_001177195.1 | 14, 15 | 1688 | ||
NM_001190267.1 | NP_001177196.1 | 14, 15 | 1686 | |||
NM_017974.3 | NP_060444.3 | 13, 14 | 1633 | |||
NM_030803.6 | NP_110430.5 | 14, 15 | 1690 | |||
NM_198890.2 | NP_942593.2 | 10, 11 | 1201 | |||
XM_005246082.2 | XP_005246139.1 | 15, 16 | 1678 | |||
XM_005246084.2 | XP_005246141.1 | 12, 13 | 1217 | |||
XM_005246086.2 | XP_005246143.1 | 11, 12 | 1166 | |||
XM_006712608.2 | XP_006712671.1 | 14, 15 | 1489 | |||
GenBank mRNA | AK001690.1 | - | 13, 14 | 1478 | ||
AK024453.1 | - | 12, 13 | 2501 | |||
AK026330.1 | - | 1, 2 | 168 | |||
AK027854.1 | - | 12, 13 | 1215 | |||
AK074434.1 | - | 8, 9 | 660 | |||
AK123876.1 | - | 11, 12 | 1052 | |||
AK308400.1 | - | 13, 14 | 1529 | |||
AY358182.1 | - | 13, 14 | 1852 | |||
AY398617.1 | - | 14, 15 | 1579 | |||
BC071846.1 | - | 10, 11 | 1039 | |||
BC117337.1 | - | 14, 15 | 1352 | |||
BC143900.1 | - | 13, 14 | 1295 | |||
EF079889.1 | - | 13, 14 | 1376 | |||
EF079890.1 | - | 10, 11 | 944 | |||
EF560734.1 | - | 13, 14 | 1503 | |||
JQ924062.1 | - | 13, 14 | 1376 |