Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: KIF26B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018012.3 | NP_060482.2 | 8 | 2106 | ||
GenBank mRNA | AK293964.1 | - | 8 | 2021 | ||
AK297303.1 | - | 8 | 2102 | |||
AY923834.1 | - | 8 | 2107 | |||
BC035896.1 | - | 5 | 627 |