Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: CHN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025201.3 | NP_001020372.2 | 11 | 1283 | ||
NM_001206602.1 | NP_001193531.1 | 5 | 1131 | |||
NM_001822.5 | NP_001813.1 | 11 | 1360 | |||
NR_038133.1 | 10 | 885 | ||||
GenBank mRNA | AB209788.1 | - | 10 | 870 | ||
AK055060.1 | - | 5 | 1062 | |||
AK289941.1 | - | 11 | 1290 | |||
AK300890.1 | - | 11 | 1277 | |||
BC011393.1 | - | 11 | 1332 | 3871858 | ||
CR456864.1 | - | 5 | 539 | |||
X51408.1 | - | 1091 | ||||
Z22641.1 | - | 11 | 1029 |