Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: RFWD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018124.3 | NP_060594.3 | 9 | 1538 | ||
XM_005256021.4 | XP_005256078.1 | 10 | 1671 | |||
XM_005256022.4 | XP_005256079.1 | 9 | 2421 | |||
XM_006721228.3 | XP_006721291.1 | 10 | 1671 | |||
XM_011523191.2 | XP_011521493.1 | 9 | 1491 | |||
XM_017023391.1 | XP_016878880.1 | 10 | 1587 | |||
XM_017023392.1 | XP_016878881.1 | 9 | 1535 | |||
GenBank mRNA | AK001382.1 | - | 4 | 440 | ||
AK291200.1 | - | 10 | 1638 | |||
AK315786.1 | - | 6 | 682 | |||
BC002574.2 | - | 7 | 915 | |||
BC059371.1 | - | 9 | 1487 | 5266451 |