Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: KIAA1551 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018169.3 | NP_060639.3 | 4 | 567 | ||
XM_005253405.2 | XP_005253462.1 | 3 | 1018 | |||
XM_006719099.2 | XP_006719162.1 | 4 | 1186 | |||
XM_011520722.1 | XP_011519024.1 | 4 | 727 | |||
XM_017019550.1 | XP_016875039.1 | 3 | 1427 | |||
GenBank mRNA | AK092399.1 | - | 4 | 537 | ||
BC140819.1 | - | 4 | 518 | 9021687 |