Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Western Blot Products
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Food and Beverage
Lab Solutions
Pharma and Biopharma
Real-Time PCR
Semiconductor Analysis
Clinical and Diagnostics
Digital Solutions
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
See all services
Help and Support
Order Help
Digital Solutions
Product Support
Technical Information
Training and Education
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other YY1AP1 SIRNA Assays ›
Gene Symbol:
YY1AP1
Gene Name:
YY1 associated protein 1
Gene Aliases:
HCCA1, HCCA2, YAP, YY1AP
Chromosome Location:
Chr.1: 155659442 - 155689032 on Build GRCh38
UniGene ID:
Hs.584927
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s30548
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
5,366.00
Your Price
Online offer:
5366.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: YY1AP1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001198899.1
NP_001185828.1
3, 4
699
NM_001198900.1
NP_001185829.1
5, 6
632
NM_001198901.1
NP_001185830.1
5, 6
522
NM_001198902.1
NP_001185831.1
5, 6
489
NM_001198903.1
NP_001185832.1
5, 6
1009
NM_001198904.1
NP_001185833.1
5, 6
1009
NM_001198905.1
NP_001185834.1
6, 7
673
NM_001198906.1
NP_001185835.1
6, 7
657
NM_018253.3
NP_060723.2
7, 8
754
NM_139118.2
NP_620829.1
6, 7
657
NM_139119.2
NP_620830.1
6, 7
670
NM_139121.2
NP_620832.1
6, 7
623
GenBank mRNA
AF203474.1
-
5, 6
689
AF206328.1
-
6, 7
589
AF466401.1
-
5, 6
463
AK001737.1
-
2, 3
188
AK023976.1
-
6, 7
625
AK225883.1
-
2, 3
239
AK297562.1
-
5, 6
822
AK298904.1
-
6, 7
653
AK303046.1
-
5, 6
672
AK303386.1
-
3, 4
677
AK309701.1
-
5, 6
828
AK315049.1
-
6, 7
704
AY604179.1
-
5, 6
494
BC001655.2
-
7, 8
684
2961648
BC001843.2
-
7, 8
684
2961648
BC003500.1
-
6, 7
623
BC008766.2
-
5, 6
566
3051391
BC009358.2
-
2, 3
250
BC014906.2
-
6, 7
602
3922542
BC044887.1
-
5, 6
629
5276742
BG325066.1
-
5, 6
490
DC300265.1
-
5, 6
522
Back To Top
More Information
Panther Classification:
Molecular Function -
transcription cofactor
gene-specific transcriptional regulator
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
regulation of transcription, DNA-templated
regulation of cell cycle
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0