Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture and Transfection
Chemicals
Chromatography
Electron Microscopes
Lab Plasticware and Supplies
Lab Centrifuges
Lab Solutions
Mass Spectrometers
Next Generation Sequencers
See all product categories
Applications
Brands
Industrial and Applied Sciences
Food and Beverage
Forensics
Lab Solutions
Life Sciences
Pharma and Biopharma
Biotechnology
Clinical and Diagnostics
Digital Solutions
See all applications
Services
Lab Instrument and Equipment Services
Enterprise Services
Custom Services
Training Services
Financial and Leasing Services
Enterprise Level Lab Informatics
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Cell Biology Services
Food Safety Inspection Services
See all services
Help and Support
Contact Us
Product Documentation
Knowledge Base and Product FAQs
Learning Centers
Supply Center
eProcurement Solutions
Lab Instrument and Equipment Support
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other FGGY SIRNA Assays ›
Gene Symbol:
FGGY
Gene Name:
FGGY carbohydrate kinase domain containing
Gene Aliases:
-
Chromosome Location:
Chr.1: 59296405 - 59762730 on Build GRCh38
UniGene ID:
Hs.444301
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s30615
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
5,366.00
Your Price
Online offer:
5366.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: FGGY
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001113411.1
NP_001106882.1
11
1326
NM_001244714.1
NP_001231643.1
9
963
NM_001278224.1
NP_001265153.1
5
482
NM_018291.3
NP_060761.3
11
1326
NR_103473.1
12
1284
XM_011541730.1
XP_011540032.1
11
1250
XM_011541731.1
XP_011540033.1
11
1184
XM_011541732.1
XP_011540034.1
11
1227
XM_011541733.1
XP_011540035.1
11
1145
XM_011541735.1
XP_011540037.1
11
1145
XM_011541736.2
XP_011540038.1
9
1091
XM_017001643.1
XP_016857132.1
11
1356
XM_017001644.1
XP_016857133.1
11
1149
XM_017001645.1
XP_016857134.1
11
1250
XM_017001646.1
XP_016857135.1
11
1227
XM_017001647.1
XP_016857136.1
11
1355
XM_017001648.1
XP_016857137.1
12
1169
XM_017001649.1
XP_016857138.1
11
1295
XM_017001650.1
XP_016857139.1
13
1518
XM_017001651.1
XP_016857140.1
13
1374
XM_017001652.1
XP_016857141.1
11
1295
XM_017001653.1
XP_016857142.1
11
1227
XM_017001654.1
XP_016857143.1
9
1046
XM_017001655.1
XP_016857144.1
9
955
XM_017001656.1
XP_016857145.1
12
1362
XM_017001657.1
XP_016857146.1
11
1150
XM_017001658.1
XP_016857147.1
12
1293
XM_017001659.1
XP_016857148.1
10
1183
XM_017001660.1
XP_016857149.1
10
1086
XM_017001661.1
XP_016857150.1
10
1243
XM_017001662.1
XP_016857151.1
10
928
XM_017001663.1
XP_016857152.1
10
1103
XM_017001664.1
XP_016857153.1
9
980
XM_017001665.1
XP_016857154.1
9
887
XM_017001666.1
XP_016857155.1
9
1041
XM_017001667.1
XP_016857156.1
9
1095
XM_017001668.1
XP_016857157.1
9
851
XM_017001669.1
XP_016857158.1
9
946
XM_017001670.1
XP_016857159.1
9
1719
XM_017001671.1
XP_016857160.1
9
847
XM_017001672.1
XP_016857161.1
10
1183
XM_017001673.1
XP_016857162.1
10
1240
XM_017001674.1
XP_016857163.1
10
1099
XM_017001675.1
XP_016857164.1
10
1183
XM_017001676.1
XP_016857165.1
10
1243
XM_017001677.1
XP_016857166.1
7
3421
XM_017001678.1
XP_016857167.1
7
4696
GenBank mRNA
AK001848.1
-
5
356
AK054842.1
-
9
954
AK090568.1
-
12
1284
AK315649.1
-
11
1211
BC000610.2
-
5
457
BC014947.1
-
11
1211
4871664
BX647626.1
-
11
1298
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_001013932, XM_008763858, XM_008763859, XM_008763857, XM_008763856, and others.
This siRNA also targets the following transcripts in Mouse: NM_001113412, AK009249, AK082737, AK138439.
Panther Classification:
Molecular Function -
carbohydrate kinase
Gene Ontology Categories:
Function(s)
Process(es)
carbohydrate metabolic process
phosphorylation
neuron cellular homeostasis
molecular_function
kinase activity
phosphotransferase activity, alcohol group as acceptor
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-55658555d6-7l2qm:80/100.66.75.127:80.
git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.41.0-Offline