Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture and Transfection
    • Chemicals
    • Chromatography
    • Electron Microscopes
    • Lab Plasticware and Supplies
    • Lab Centrifuges
    • Lab Solutions
    • Mass Spectrometers
    • Next Generation Sequencers
    • See all product categories
  • Applications
    • Brands
    • Industrial and Applied Sciences
    • Food and Beverage
    • Forensics
    • Lab Solutions
    • Life Sciences
    • Pharma and Biopharma
    • Biotechnology
    • Clinical and Diagnostics
    • Digital Solutions
    • See all applications
  • Services
    • Lab Instrument and Equipment Services
    • Custom Services
    • Training Services
    • Financial and Leasing Services
    • Enterprise Level Lab Informatics
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Cell Biology Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • Contact Us
    • Product Documentation
    • Knowledge Base and Product FAQs
    • Learning Centers
    • Supply Center
    • eProcurement Solutions
    • Lab Instrument and Equipment Support
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other PPP6R3 SIRNA Assays ›
          Gene Symbol:
          PPP6R3
          Gene Name:
          protein phosphatase 6 regulatory subunit 3
          Gene Aliases:
          C11orf23, PP6R3, SAP190, SAPL, SAPLa, SAPS3
          Chromosome Location:
          Chr.11: 68460712 - 68615334 on Build GRCh38
          UniGene ID:
          Hs.503022
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s30659

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: PPP6R3
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001164160.1 NP_001157632.1 16 1917
          NM_001164161.1 NP_001157633.1 16 1917
          NM_001164163.1 NP_001157635.1 15 1830
          NM_001164164.1 NP_001157636.1 15 1782
          NM_018312.4 NP_060782.2 15 1782
          XM_006718608.3 XP_006718671.1 17 2033
          XM_006718613.2 XP_006718676.1 17 2033
          XM_006718618.3 XP_006718681.1 16 1946
          XM_006718622.3 XP_006718685.1 15 1793
          XM_006718623.3 XP_006718686.1 15 1776
          XM_006718624.2 XP_006718687.1 15 1793
          XM_011545135.1 XP_011543437.1 17 2033
          XM_011545136.2 XP_011543438.1 16 1916
          XM_011545137.2 XP_011543439.1 17 2016
          XM_011545138.2 XP_011543440.1 18 2234
          XM_011545140.2 XP_011543442.1 17 2033
          XM_011545142.2 XP_011543444.1 16 1946
          XM_011545145.2 XP_011543447.1 16 1880
          XM_017017962.1 XP_016873451.1 16 1871
          XM_017017963.1 XP_016873452.1 17 1957
          XM_017017971.1 XP_016873460.1 16 1946
          XM_017017972.1 XP_016873461.1 15 1802
          XM_017017973.1 XP_016873462.1 16 1929
          XM_017017977.1 XP_016873466.1 16 1804
          XM_017017978.1 XP_016873467.1 15 1655
          XM_017017980.1 XP_016873469.1 15 1655
          XM_017017982.1 XP_016873471.1 14 1568
          XM_017017983.1 XP_016873472.1 16 2054
          XM_017017984.1 XP_016873473.1 15 1897
          GenBank mRNA AB046778.2 - 16 1865
          AF264779.1 - 14 1687
          AF264780.1 - 14 1687
          AK057250.1 - 10 990
          AK125055.1 - 1 1669
          AK300359.1 - 8 751
          AK302611.1 - 8 771
          AK303315.1 - 11 1233
          AK315136.1 - 15 1799
          BC105933.1 - 15 1785 40029064
          BC105934.1 - 15 1785 40029058
          BC107599.1 - 14 1698 40029065
          CR933658.1 - 16 1880
          CR936718.1 - 16 1885
          DQ111954.2 - 14 1650

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_001287134, XM_008760122, XM_008760125, XM_008760130, XM_008760121, and others.
          This siRNA also targets the following transcripts in Mouse: NM_029456, NM_028999, BC008529, AK035236, CF727136.

          Panther Classification:

          Molecular Function -

          phosphatase modulator

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of phosphoprotein phosphatase activity
          COPII vesicle coating
          protein binding
          protein phosphatase binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Quick Order
          • eProcurement
          • Supply Center
          • Order Status
          • Chemicals
          • India Mobile App
          • Government eMarketplace
          Support Plus Icon Minus Icon
          • Order Support
          • Training
          • Contact Us
          • Report a Site Issue
          • Instrument Management
          Resources Plus Icon Minus Icon
          • Product Selection Guides
          • Mobile & Desktop Apps
          • Webinars
          • Blog 
          • Social Media
          • New Products
          • Promotions
          • Shared Lists
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          India flag icon
          India

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-vnb2r:80/100.66.79.197:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0