Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture and Transfection
    • Chemicals
    • Chromatography
    • Electron Microscopes
    • Lab Plasticware and Supplies
    • Lab Centrifuges
    • Lab Solutions
    • Mass Spectrometers
    • Next Generation Sequencers
    • See all product categories
  • Applications
    • Brands
    • Industrial and Applied Sciences
    • Food and Beverage
    • Forensics
    • Lab Solutions
    • Life Sciences
    • Pharma and Biopharma
    • Biotechnology
    • Clinical and Diagnostics
    • Digital Solutions
    • See all applications
  • Services
    • Lab Instrument and Equipment Services
    • Enterprise Services
    • Custom Services
    • Training Services
    • Financial and Leasing Services
    • Enterprise Level Lab Informatics
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Cell Biology Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • Contact Us
    • Product Documentation
    • Knowledge Base and Product FAQs
    • Learning Centers
    • Supply Center
    • eProcurement Solutions
    • Lab Instrument and Equipment Support
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other FBXW7 SIRNA Assays ›
          Gene Symbol:
          FBXW7
          Gene Name:
          F-box and WD repeat domain containing 7
          Gene Aliases:
          AGO, CDC4, FBW6, FBW7, FBX30, FBXO30, FBXW6, SEL-10, SEL10, hAgo, hCdc4
          Chromosome Location:
          Chr.4: 152321258 - 152536095 on Build GRCh38
          UniGene ID:
          Hs.561245
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s30663

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: FBXW7
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001013415.1 NP_001013433.1 9 1383
          NM_018315.4 NP_060785.2 9 1548
          NM_033632.3 NP_361014.1 10 1722
          XM_011532083.1 XP_011530385.1 12 2735
          XM_011532084.1 XP_011530386.1 11 2685
          XM_011532085.1 XP_011530387.1 10 2592
          XM_011532086.1 XP_011530388.1 13 2913
          XM_011532087.1 XP_011530389.1 12 2863
          XM_011532088.1 XP_011530390.1 10 2121
          XM_017008362.1 XP_016863851.1 12 3132
          GenBank mRNA AB209725.1 - 9 1312
          AF383178.1 - 9 1122
          AF411971.1 - 9 1635
          AF411972.1 - 9 1416
          AK001933.1 - 9 1173
          AK131383.1 - 2 1008
          AK223449.1 - 9 1350
          AK294568.1 - 8 1109
          AK299722.1 - 8 1131
          AK315142.1 - 9 1330
          AY008274.1 - 9 1385
          AY033553.1 - 9 1416
          AY049984.1 - 9 1629
          BC037320.2 - 9 1303
          BC117244.1 - 10 1690 40125795
          BC117246.1 - 10 1690 40125797
          BC143944.1 - 11 1740 9052462
          CR749305.1 - 10 1853

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          protein polyubiquitination
          vasculature development
          sister chromatid cohesion
          negative regulation of triglyceride biosynthetic process
          regulation of lipid storage
          viral process
          protein ubiquitination
          SCF-dependent proteasomal ubiquitin-dependent protein catabolic process
          positive regulation of protein ubiquitination
          negative regulation of DNA endoreduplication
          regulation of protein localization
          positive regulation of epidermal growth factor-activated receptor activity
          negative regulation of Notch signaling pathway
          protein stabilization
          positive regulation of ubiquitin-protein transferase activity
          lipid homeostasis
          positive regulation of ERK1 and ERK2 cascade
          positive regulation of proteasomal protein catabolic process
          regulation of cell cycle G1/S phase transition
          regulation of mitophagy
          positive regulation of oxidative stress-induced neuron intrinsic apoptotic signaling pathway
          positive regulation of protein targeting to mitochondrion
          positive regulation of protein ubiquitination involved in ubiquitin-dependent protein catabolic process
          negative regulation of hepatocyte proliferation
          negative regulation of SREBP signaling pathway
          ubiquitin-protein transferase activity
          protein binding
          cyclin binding
          protein binding, bridging
          ubiquitin protein ligase binding
          identical protein binding
          phosphothreonine binding
          ubiquitin-protein transferase activator activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Quick Order
          • eProcurement
          • Supply Center
          • Order Status
          • Chemicals
          • India Mobile App
          • Government eMarketplace
          Support Plus Icon Minus Icon
          • Order Support
          • Training
          • Contact Us
          • Report a Site Issue
          • Instrument Management
          Resources Plus Icon Minus Icon
          • Product Selection Guides
          • Mobile & Desktop Apps
          • Webinars
          • Blog 
          • Social Media
          • New Products
          • Promotions
          • Shared Lists
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          India flag icon
          India

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-lmjtd:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0