Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other FBXW7 SIRNA Assays ›
          Gene Symbol:
          FBXW7
          Gene Name:
          F-box and WD repeat domain containing 7
          Gene Aliases:
          AGO, CDC4, FBW6, FBW7, FBX30, FBXO30, FBXW6, SEL-10, SEL10, hAgo, hCdc4
          Chromosome Location:
          Chr.4: 152321258 - 152536095 on Build GRCh38
          UniGene ID:
          Hs.561245
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s30665

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price 5,366.00
          Your Price
          Online offer:
          5366.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: FBXW7
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001013415.1 NP_001013433.1 11 1789
          NM_018315.4 NP_060785.2 11 1954
          NM_033632.3 NP_361014.1 12 2128
          XM_011532083.1 XP_011530385.1 14 3141
          XM_011532084.1 XP_011530386.1 13 3091
          XM_011532085.1 XP_011530387.1 12 2998
          XM_011532086.1 XP_011530388.1 15 3319
          XM_011532087.1 XP_011530389.1 14 3269
          XM_011532088.1 XP_011530390.1 12 2527
          XM_017008362.1 XP_016863851.1 14 3538
          GenBank mRNA AB209725.1 - 11 1718
          AF383178.1 - 11 1528
          AF411971.1 - 11 2041
          AF411972.1 - 11 1822
          AK001933.1 - 11 1579
          AK131383.1 - 2 4059
          AK223449.1 - 11 1756
          AK294568.1 - 10 1515
          AK299722.1 - 10 1537
          AK315142.1 - 11 1735
          AY008274.1 - 11 1791
          AY033553.1 - 11 1822
          AY049984.1 - 11 2035
          BC037320.2 - 11 1709
          BC117244.1 - 12 2096 40125795
          BC117246.1 - 12 2096 40125797
          BC143944.1 - 13 2146 9052462
          CR749305.1 - 12 2259

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          protein polyubiquitination
          vasculature development
          sister chromatid cohesion
          negative regulation of triglyceride biosynthetic process
          regulation of lipid storage
          viral process
          protein ubiquitination
          SCF-dependent proteasomal ubiquitin-dependent protein catabolic process
          positive regulation of protein ubiquitination
          negative regulation of DNA endoreduplication
          regulation of protein localization
          positive regulation of epidermal growth factor-activated receptor activity
          negative regulation of Notch signaling pathway
          protein stabilization
          positive regulation of ubiquitin-protein transferase activity
          lipid homeostasis
          positive regulation of ERK1 and ERK2 cascade
          positive regulation of proteasomal protein catabolic process
          regulation of cell cycle G1/S phase transition
          regulation of mitophagy
          positive regulation of oxidative stress-induced neuron intrinsic apoptotic signaling pathway
          positive regulation of protein targeting to mitochondrion
          positive regulation of protein ubiquitination involved in ubiquitin-dependent protein catabolic process
          negative regulation of hepatocyte proliferation
          negative regulation of SREBP signaling pathway
          ubiquitin-protein transferase activity
          protein binding
          cyclin binding
          protein binding, bridging
          ubiquitin protein ligase binding
          identical protein binding
          phosphothreonine binding
          ubiquitin-protein transferase activator activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0