Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Instrument Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
Our Instagram
Our Facebook
Blog Behind the Bench
Customer Experience Center (CEC)
Ecommerce Exclusives
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PPP1R9A SIRNA Assays ›
Gene Symbol:
PPP1R9A
Gene Name:
protein phosphatase 1 regulatory subunit 9A
Gene Aliases:
NRB1, NRBI, Neurabin-I
Chromosome Location:
Chr.7: 94907202 - 95296415 on Build GRCh38
UniGene ID:
Hs.21816
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s31059
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
5,366.00
Your Price
Online offer:
5366.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PPP1R9A
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001166160.1
NP_001159632.1
4, 5
1936
NM_001166161.1
NP_001159633.1
3, 4
1870
NM_001166162.1
NP_001159634.1
3, 4
1870
NM_001166163.1
NP_001159635.1
3, 4
1870
NM_017650.2
NP_060120.2
4, 5
1936
XM_011516380.1
XP_011514682.1
4, 5
2060
XM_011516381.1
XP_011514683.1
4, 5
1936
XM_011516382.2
XP_011514684.1
4, 5
1949
XM_011516383.1
XP_011514685.1
4, 5
1990
XM_011516389.1
XP_011514691.1
4, 5
1936
XM_017012394.1
XP_016867883.1
4, 5
1936
XM_017012395.1
XP_016867884.1
4, 5
1936
XM_017012396.1
XP_016867885.1
4, 5
2060
XM_017012397.1
XP_016867886.1
4, 5
1936
XM_017012398.1
XP_016867887.1
4, 5
1948
XM_017012399.1
XP_016867888.1
4, 5
1936
XM_017012400.1
XP_016867889.1
4, 5
1936
XM_017012401.1
XP_016867890.1
4, 5
1936
XM_017012402.1
XP_016867891.1
4, 5
1936
XM_017012403.1
XP_016867892.1
4, 5
1936
XM_017012404.1
XP_016867893.1
4, 5
1936
XM_017012405.1
XP_016867894.1
4, 5
2060
XM_017012406.1
XP_016867895.1
4, 5
1991
XM_017012407.1
XP_016867896.1
4, 5
1936
XM_017012409.1
XP_016867898.1
4, 5
1936
XM_017012410.1
XP_016867899.1
4, 5
1980
GenBank mRNA
AB033048.1
-
3, 4
588
AK096514.1
-
4, 5
1899
BC130449.1
-
3, 4
1685
40146480
BC144108.1
-
3, 4
1686
BC144109.1
-
3, 4
1685
9052629
BC150636.1
-
3, 4
1884
9057006
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
actin filament organization
aging
calcium-mediated signaling
regulation of actin filament polymerization
neuron projection development
positive regulation of protein kinase activity
regulation of filopodium assembly
negative regulation of stress fiber assembly
regulation of synapse structural plasticity
regulation of synapse assembly
excitatory postsynaptic potential
positive regulation of dendritic spine development
regulation of dendritic spine morphogenesis
cellular response to toxic substance
negative regulation of long-term synaptic potentiation
positive regulation of long term synaptic depression
negative regulation of spontaneous neurotransmitter secretion
protein binding
protein C-terminus binding
protein phosphatase 1 binding
protein kinase binding
protein domain specific binding
protein homodimerization activity
ion channel binding
actin filament binding
GTPase binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0