Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: LIMS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136037.2 | NP_001129509.2 | 11 | 1758 | ||
NM_001161403.1 | NP_001154875.1 | 10 | 1692 | |||
NM_001161404.1 | NP_001154876.1 | 10 | 1982 | |||
NM_001256542.1 | NP_001243471.1 | 7 | 1384 | |||
NM_017980.4 | NP_060450.2 | 10 | 1803 | |||
XM_005263710.2 | XP_005263767.1 | 9 | 1475 | |||
XM_006712627.3 | XP_006712690.1 | 7 | 1284 | |||
XM_011511453.1 | XP_011509755.1 | 9 | 1539 | |||
XM_017004469.1 | XP_016859958.1 | 5 | 1249 | |||
GenBank mRNA | AF520987.1 | - | 10 | 2145 | ||
AF527764.1 | - | 10 | 1757 | |||
AF527765.1 | - | 10 | 1619 | |||
AF527766.1 | - | 10 | 1659 | |||
AF527767.1 | - | 9 | 1915 | |||
AF527768.1 | - | 8 | 1274 | |||
AF527769.1 | - | 6 | 1348 | |||
AF527770.1 | - | 7 | 1311 | |||
AK000906.1 | - | 8 | 1174 | |||
AK022470.1 | - | 10 | 1571 | |||
AK055363.1 | - | 10 | 2147 | |||
AK091652.1 | - | 8 | 2053 | |||
AK093692.1 | - | 4 | 1746 | |||
AK094954.1 | - | 9 | 2239 | |||
AK095790.1 | - | 13 | 2207 | |||
AK123014.1 | - | 5 | 4451 | |||
AL512716.1 | - | 1 | 631 | |||
BC001370.2 | - | 2 | 782 | |||
BC008977.1 | - | 2 | 697 | |||
BC065816.1 | - | 10 | 1610 | 6181259 |