Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other KANSL3 SIRNA Assays ›
Gene Symbol:
KANSL3
Gene Name:
KAT8 regulatory NSL complex subunit 3
Gene Aliases:
KIAA1310, NSL3, Rcd1
Chromosome Location:
Chr.2: 96593155 - 96638379 on Build GRCh38
UniGene ID:
Hs.516341
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s31242
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
5,366.00
Your Price
Online offer:
5366.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: KANSL3
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001115016.2
NP_001108488.1
15
2011
NR_047653.1
15
2072
NR_047654.1
15
2072
NR_047655.1
15
1998
NR_047656.1
14
1920
NR_047657.1
16
2089
NR_047658.1
15
1988
XM_005263982.2
XP_005264039.1
16
2088
XM_005263984.1
XP_005264041.1
16
2088
XM_005263987.1
XP_005264044.1
15
2010
XM_011511454.2
XP_011509756.1
16
2088
XM_011511455.2
XP_011509757.1
16
2057
XM_011511456.2
XP_011509758.1
16
2091
XM_011511457.2
XP_011509759.1
16
2010
XM_011511458.2
XP_011509760.1
16
2088
XM_011511459.2
XP_011509761.1
15
2013
XM_011511460.2
XP_011509762.1
15
2010
XM_011511461.2
XP_011509763.1
15
2010
XM_011511462.2
XP_011509764.1
15
1917
XM_011511463.2
XP_011509765.1
16
2101
XM_011511464.2
XP_011509766.1
16
2160
XM_011511466.2
XP_011509768.1
15
2023
XM_011511467.2
XP_011509769.1
15
1943
XM_017004470.1
XP_016859959.1
16
2088
XM_017004471.1
XP_016859960.1
15
1995
XM_017004472.1
XP_016859961.1
16
2101
XM_017004473.1
XP_016859962.1
16
2101
XM_017004474.1
XP_016859963.1
16
1996
GenBank mRNA
AB037731.1
-
16
2041
AF311326.1
-
17
1824
AK023813.1
-
15
2039
AK091163.1
-
1
1446
AK302646.1
-
15
1939
AK304011.1
-
14
1920
AK314367.1
-
15
1948
AL136849.1
-
1
12
AY050169.1
-
15
2039
BC024276.1
-
15
1948
BC051763.1
-
14
1720
BC063792.1
-
15
1941
6504387
BC127820.1
-
3
388
DB133508.1
-
2
183
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
histone H4-K5 acetylation
histone H4-K8 acetylation
histone H4-K16 acetylation
histone acetyltransferase activity (H4-K5 specific)
histone acetyltransferase activity (H4-K8 specific)
histone acetyltransferase activity (H4-K16 specific)
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0