Search
Search

CLCA1| Gene Symbol: CLCA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001285.3 | NP_001276.2 | 1 | 238 | ||
| GenBank mRNA | AF039400.1 | - | 2 | 460 | ||
| AF127036.1 | - | 1 | 113 | |||
| AK097057.1 | - | 1 | 153 | |||
| AK291999.1 | - | 1 | 154 | |||
| AK314375.1 | - | 1 | 156 | |||
| DA428844.1 | - | 1 | 238 | |||