Search Thermo Fisher Scientific
- ์ฃผ๋ฌธ ํํฉ
- ๋น ๋ฅธ์ฃผ๋ฌธ
-
ํ์์ด ์๋์ ๊ฐ์? ๊ณ์ ์์ฑํ๊ธฐ
Search Thermo Fisher Scientific
Gene Symbol: ARHGEF40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278529.1 | NP_001265458.1 | 13 | 2725 | ||
NM_001278530.1 | NP_001265459.1 | 13 | 2531 | |||
NM_018071.4 | NP_060541.3 | 13 | 2698 | |||
XM_005267844.2 | XP_005267901.1 | 13 | 2734 | |||
XM_011536937.2 | XP_011535239.1 | 13 | 2734 | |||
XM_011536938.2 | XP_011535240.1 | 13 | 2736 | |||
XM_017021434.1 | XP_016876923.1 | 14 | 2991 | |||
XM_017021435.1 | XP_016876924.1 | 12 | 3208 | |||
XM_017021436.1 | XP_016876925.1 | 9 | 991 | |||
XM_017021437.1 | XP_016876926.1 | 9 | 991 | |||
GenBank mRNA | AK024463.1 | - | 11 | 1936 | ||
AK074057.1 | - | 13 | 2643 | |||
AK126755.1 | - | 9 | 793 | |||
AL137291.1 | - | 1 | 41 | |||
BC142692.1 | - | 13 | 2668 | 40148708 | ||
BC151212.1 | - | 13 | 2668 |