Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other COA1 SIRNA Assays ›
Gene Symbol:
COA1
Gene Name:
cytochrome c oxidase assembly factor 1 homolog
Gene Aliases:
C7orf44, MITRAC15
Chromosome Location:
Chr.7: 43608452 - 43729541 on Build GRCh38
UniGene ID:
Hs.268887
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s31395
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
5,366.00
Your Price
Online offer:
5366.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: COA1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001321197.1
NP_001308126.1
4, 5
496
NM_001321198.1
NP_001308127.1
4, 5
377
NM_001321199.1
NP_001308128.1
4, 5
341
NM_001321200.1
NP_001308129.1
4, 5
397
NM_001321201.1
NP_001308130.1
3, 4
269
NM_018224.3
NP_060694.2
3, 4
276
NR_047701.1
4, 5
341
NR_135580.1
4, 5
377
NR_135581.1
4, 5
377
NR_135582.1
3, 4
276
NR_135583.1
3, 4
276
XM_005249807.4
XP_005249864.1
3, 4
340
XM_006715752.2
XP_006715815.1
4, 5
308
XM_017012413.1
XP_016867902.1
3, 4
267
XM_017012414.1
XP_016867903.1
3, 4
267
XM_017012415.1
XP_016867904.1
4, 5
369
XM_017012416.1
XP_016867905.1
3, 4
267
XM_017012417.1
XP_016867906.1
3, 4
267
XM_017012418.1
XP_016867907.1
3, 4
267
XM_017012419.1
XP_016867908.1
3, 4
267
XM_017012420.1
XP_016867909.1
4, 5
369
XM_017012421.1
XP_016867910.1
4, 5
491
XM_017012422.1
XP_016867911.1
4, 5
380
XM_017012423.1
XP_016867912.1
4, 5
300
XM_017012424.1
XP_016867913.1
3, 4
283
XM_017012425.1
XP_016867914.1
4, 5
384
XM_017012426.1
XP_016867915.1
4, 5
503
XM_017012427.1
XP_016867916.1
5, 6
458
XM_017012428.1
XP_016867917.1
3, 4
283
XM_017012429.1
XP_016867918.1
2, 3
177
XM_017012430.1
XP_016867919.1
4, 5
369
XM_017012431.1
XP_016867920.1
3, 4
283
XM_017012432.1
XP_016867921.1
3, 4
283
GenBank mRNA
AK001665.1
-
4, 5
310
AK021905.1
-
4, 5
446
AK023007.1
-
4, 5
293
AK023053.1
-
4, 5
309
AK097203.1
-
2, 3
1807
AK293043.1
-
4, 5
319
BC056884.1
-
4, 5
268
6159045
BG028456.1
-
298
BG334368.1
-
4, 5
282
BG548154.1
-
4, 5
333
BM918741.1
-
3, 4
249
BP373150.1
-
5, 6
514
BP377872.1
-
3, 4
277
BQ231567.1
-
3, 4
214
DA742258.1
-
3, 4
258
Back To Top
More Information
Panther Classification:
Molecular Function -
chaperone
Gene Ontology Categories:
Function(s)
Process(es)
translation
mitochondrial respiratory chain complex I assembly
mitochondrial respiratory chain complex IV assembly
RNA binding
structural constituent of ribosome
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-mtlvn:80/100.66.79.31:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0