Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other COA1 SIRNA Assays ›
          Gene Symbol:
          COA1
          Gene Name:
          cytochrome c oxidase assembly factor 1 homolog
          Gene Aliases:
          C7orf44, MITRAC15
          Chromosome Location:
          Chr.7: 43608452 - 43729541 on Build GRCh38
          UniGene ID:
          Hs.268887
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s31396

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price 5,366.00
          Your Price
          Online offer:
          5366.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: COA1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001321197.1 NP_001308126.1 6 652
          NM_001321198.1 NP_001308127.1 6 533
          NM_001321199.1 NP_001308128.1 6 497
          NM_001321200.1 NP_001308129.1 6 553
          NM_001321201.1 NP_001308130.1 5 425
          NM_018224.3 NP_060694.2 5 432
          NR_047701.1 6 497
          NR_135580.1 6 533
          NR_135581.1 6 533
          NR_135582.1 5 432
          NR_135583.1 5 432
          XM_005249807.4 XP_005249864.1 5 496
          XM_006715752.2 XP_006715815.1 6 464
          XM_017012413.1 XP_016867902.1 5 423
          XM_017012414.1 XP_016867903.1 5 423
          XM_017012415.1 XP_016867904.1 6 525
          XM_017012416.1 XP_016867905.1 5 423
          XM_017012417.1 XP_016867906.1 5 423
          XM_017012418.1 XP_016867907.1 5 423
          XM_017012419.1 XP_016867908.1 5 423
          XM_017012420.1 XP_016867909.1 6 525
          XM_017012421.1 XP_016867910.1 6 647
          XM_017012422.1 XP_016867911.1 6 536
          XM_017012423.1 XP_016867912.1 6 456
          XM_017012424.1 XP_016867913.1 5 439
          XM_017012425.1 XP_016867914.1 6 540
          XM_017012426.1 XP_016867915.1 6 659
          XM_017012427.1 XP_016867916.1 7 614
          XM_017012428.1 XP_016867917.1 5 439
          XM_017012429.1 XP_016867918.1 4 333
          XM_017012430.1 XP_016867919.1 6 525
          XM_017012431.1 XP_016867920.1 5 439
          XM_017012432.1 XP_016867921.1 5 439
          GenBank mRNA AK001665.1 - 6 466
          AK021905.1 - 6 602
          AK023007.1 - 6 449
          AK023053.1 - 6 465
          AK097203.1 - 4 1963
          AK129690.1 - 2 1156
          AK293043.1 - 6 475
          BC056884.1 - 6 424 6159045
          BG028456.1 - 454
          BG334368.1 - 6 442
          BG548154.1 - 6 489
          BM918741.1 - 5 405
          BQ231567.1 - 5 370
          DA742258.1 - 5 414

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          chaperone

          Gene Ontology Categories:

          Function(s) Process(es)

          translation
          mitochondrial respiratory chain complex I assembly
          mitochondrial respiratory chain complex IV assembly
          RNA binding
          structural constituent of ribosome

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0