Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: CNDP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168499.1 | NP_001161971.1 | 6 | 1128 | ||
NM_018235.2 | NP_060705.2 | 9 | 1253 | |||
XM_005266728.2 | XP_005266785.1 | 9 | 1189 | |||
XM_006722503.2 | XP_006722566.1 | 9 | 1050 | |||
XM_011526071.2 | XP_011524373.1 | 9 | 1118 | |||
XM_011526072.2 | XP_011524374.1 | 9 | 2073 | |||
XM_011526073.2 | XP_011524375.1 | 8 | 1306 | |||
GenBank mRNA | AF258592.1 | - | 6 | 856 | ||
AJ347717.1 | - | 8 | 1017 | |||
AK001262.1 | - | 4 | 518 | |||
AK001692.1 | - | 9 | 1156 | |||
AK001983.1 | - | 4 | 433 | |||
AK024471.1 | - | 5 | 622 | |||
AK097155.1 | - | 9 | 1268 | |||
AK298309.1 | - | 9 | 872 | |||
AK300904.1 | - | 9 | 1120 | |||
BC001375.2 | - | 9 | 1126 | 3051369 | ||
BC003176.1 | - | 8 | 1050 | 2957870 | ||
EU794600.1 | - | 8 | 1050 |