Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: UGGT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020121.3 | NP_064506.3 | 33 | 3940 | ||
XM_011521094.1 | XP_011519396.1 | 34 | 3976 | |||
XM_011521095.1 | XP_011519397.1 | 33 | 3895 | |||
XM_011521096.1 | XP_011519398.1 | 33 | 3844 | |||
XM_011521097.1 | XP_011519399.1 | 34 | 3976 | |||
XM_011521098.1 | XP_011519400.1 | 27 | 2993 | |||
XM_011521099.2 | XP_011519401.1 | 26 | 2910 | |||
XM_011521102.2 | XP_011519404.1 | 24 | 2684 | |||
XM_011521103.2 | XP_011519405.1 | 24 | 2742 | |||
XM_017020662.1 | XP_016876151.1 | 32 | 3763 | |||
XM_017020663.1 | XP_016876152.1 | 34 | 3878 | |||
GenBank mRNA | AF227906.2 | - | 33 | 3841 | ||
AL133051.1 | - | 30 | 3320 | |||
BC125233.1 | - | 33 | 3796 | 40124676 |