Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: UGGT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020121.3 | NP_064506.3 | 19, 20 | 2371 | ||
XM_011521094.1 | XP_011519396.1 | 19, 20 | 2326 | |||
XM_011521095.1 | XP_011519397.1 | 19, 20 | 2326 | |||
XM_011521096.1 | XP_011519398.1 | 19, 20 | 2326 | |||
XM_011521097.1 | XP_011519399.1 | 19, 20 | 2326 | |||
XM_011521098.1 | XP_011519400.1 | 12, 13 | 1343 | |||
XM_011521099.2 | XP_011519401.1 | 11, 12 | 1260 | |||
XM_011521100.2 | XP_011519402.1 | 19, 20 | 2326 | |||
XM_011521102.2 | XP_011519404.1 | 9, 10 | 1034 | |||
XM_011521103.2 | XP_011519405.1 | 9, 10 | 1092 | |||
XM_017020662.1 | XP_016876151.1 | 19, 20 | 2326 | |||
XM_017020663.1 | XP_016876152.1 | 20, 21 | 2309 | |||
GenBank mRNA | AF227906.2 | - | 19, 20 | 2272 | ||
AK001735.1 | - | 19, 20 | 2296 | |||
AL133051.1 | - | 16, 17 | 1751 | |||
BC125233.1 | - | 19, 20 | 2227 | 40124676 |