Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Western Blot Products
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Food and Beverage
Lab Solutions
Pharma and Biopharma
Real-Time PCR
Semiconductor Analysis
Clinical and Diagnostics
Digital Solutions
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
See all services
Help and Support
Order Help
Digital Solutions
Product Support
Technical Information
Training and Education
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SPATA7 SIRNA Assays ›
Gene Symbol:
SPATA7
Gene Name:
spermatogenesis associated 7
Gene Aliases:
HEL-S-296, HSD-3.1, HSD3, LCA3
Chromosome Location:
Chr.14: 88385638 - 88470350 on Build GRCh38
UniGene ID:
Hs.525518
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s31573
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SPATA7
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001040428.3
NP_001035518.1
10
1258
NM_018418.4
NP_060888.2
11
1354
XM_005267851.1
XP_005267908.1
11
1328
XM_005267852.1
XP_005267909.1
10
1257
XM_005267854.1
XP_005267911.1
12
1390
XM_005267855.1
XP_005267912.1
11
1323
XM_006720204.1
XP_006720267.1
11
1328
XM_006720205.1
XP_006720268.1
11
1328
XM_011536951.1
XP_011535253.1
11
1522
XM_011536952.1
XP_011535254.1
9
1140
XM_011536953.1
XP_011535255.1
10
1461
XM_017021452.1
XP_016876941.1
11
1519
XM_017021453.1
XP_016876942.1
11
1488
XM_017021454.1
XP_016876943.1
12
1387
XM_017021455.1
XP_016876944.1
11
1320
XM_017021456.1
XP_016876945.1
12
1333
XM_017021457.1
XP_016876946.1
10
1458
GenBank mRNA
AF144487.3
-
11
1334
AF144488.2
-
9
1062
AK055864.1
-
10
2926
AK225746.1
-
12
1328
AK290631.1
-
10
1229
AL136604.1
-
10
1238
BC008656.1
-
6
786
BC090875.1
-
11
1641
30383306
CR533563.1
-
10
1083
CR936777.1
-
11
1300
EU668353.1
-
11
1330
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
spermatogenesis
visual perception
photoreceptor cell maintenance
response to stimulus
protein localization to photoreceptor outer segment
protein localization to photoreceptor connecting cilium
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-567f868b4f-2g8tf:80/10.239.165.190:80.
git-commit: 96b0dd45ad55c22ec1de1c352dff25df4a817ab2
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.8.0-2024.05.19-1.0