Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: CLK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001162407.1 | NP_001155879.1 | 10 | 1585 | ||
NM_004071.3 | NP_004062.2 | 10 | 1305 | |||
NR_027855.1 | 9 | 1214 | ||||
NR_027856.1 | 8 | 2705 | ||||
GenBank mRNA | AK025306.1 | - | 5 | 2612 | ||
AK294295.1 | - | 10 | 1585 | |||
AK316127.1 | - | 10 | 1276 | |||
BC028149.1 | - | 9 | 1110 | |||
BC031549.1 | - | 10 | 1201 | 4387232 | ||
L29219.1 | - | 10 | 1279 | |||
L29222.1 | - | 9 | 1188 | |||
M59287.1 | - | 9 | 1189 |