Search
Search

CLK1| Gene Symbol: CLK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001162407.1 | NP_001155879.1 | 4 | 919 | ||
| NM_004071.3 | NP_004062.2 | 4 | 639 | |||
| NR_027856.1 | 3 | 1661 | ||||
| GenBank mRNA | AK025306.1 | - | 1 | 836 | ||
| AK294295.1 | - | 4 | 919 | |||
| AK307301.1 | - | 4 | 769 | |||
| AK316127.1 | - | 4 | 610 | |||
| BC031549.1 | - | 4 | 535 | 4387232 | ||
| L29219.1 | - | 4 | 613 | |||