Search
Search

APOM| Gene Symbol: APOM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001256169.1 | NP_001243098.1 | 5 | 550 | ||
| NM_019101.2 | NP_061974.2 | 5 | 595 | |||
| NR_045828.1 | 5 | 557 | ||||
| XM_006715150.3 | XP_006715213.1 | 5 | 579 | |||
| GenBank mRNA | AF118393.3 | - | 5 | 555 | ||
| AF161454.1 | - | 5 | 516 | |||
| BC020683.1 | - | 5 | 574 | 4733993 | ||
| BG564719.1 | - | 5 | 597 | |||
| CN428415.1 | - | 5 | 537 | |||