Search Thermo Fisher Scientific
- ์ฃผ๋ฌธ ํํฉ
- ๋น ๋ฅธ์ฃผ๋ฌธ
-
ํ์์ด ์๋์ ๊ฐ์? ๊ณ์ ์์ฑํ๊ธฐ
Search Thermo Fisher Scientific
Gene Symbol: WDR45B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019613.3 | NP_062559.2 | 2 | 308 | ||
XM_005256377.4 | XP_005256434.1 | 2 | 233 | |||
XM_017024862.1 | XP_016880351.1 | 2 | 223 | |||
XM_017024863.1 | XP_016880352.1 | 2 | 67 | |||
GenBank mRNA | AK297477.1 | - | 2 | 62 | ||
AM182326.1 | - | 2 | 113 | |||
BC000974.2 | - | 2 | 91 | |||
BC007838.1 | - | 2 | 280 | |||
CN262716.1 | - | 2 | 308 |