Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: RGMA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166283.1 | NP_001159755.1 | 3 | 826 | ||
NM_001166286.1 | NP_001159758.1 | 2 | 686 | |||
NM_001166287.1 | NP_001159759.1 | 3 | 800 | |||
NM_001166288.1 | NP_001159760.1 | 3 | 684 | |||
NM_001166289.1 | NP_001159761.1 | 3 | 704 | |||
NM_020211.2 | NP_064596.2 | 3 | 853 | |||
GenBank mRNA | AK074910.1 | - | 3 | 843 | ||
AK074966.1 | - | 2 | 660 | |||
AK074980.1 | - | 3 | 620 | |||
AK125204.1 | - | 4 | 809 | |||
AK291365.1 | - | 2 | 483 | |||
AK299363.1 | - | 3 | 702 | |||
AK309814.1 | - | 4 | 1066 | |||
AL136826.1 | - | 3 | 667 | |||
AL390083.1 | - | 2 | 609 | |||
BC015886.1 | - | 2 | 677 | |||
BC140838.1 | - | 3 | 631 | 9021706 | ||
BC151132.1 | - | 3 | 631 | 9057032 |