Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: POGLUT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152305.2 | NP_689518.1 | 2, 3 | 260 | ||
NR_024265.1 | 2, 3 | 260 | ||||
XM_006713705.3 | XP_006713768.1 | 1, 2 | 71 | |||
XM_017006878.1 | XP_016862367.1 | 1, 2 | 91 | |||
XM_017006879.1 | XP_016862368.1 | 2, 3 | 206 | |||
XM_017006880.1 | XP_016862369.1 | 2, 3 | 206 | |||
GenBank mRNA | AF168711.1 | - | 2, 3 | 211 | ||
AK075444.1 | - | 2, 3 | 196 | |||
AK222937.1 | - | 2, 3 | 220 | |||
AK295984.1 | - | 2, 3 | 235 | |||
AK301002.1 | - | 2, 3 | 235 | |||
AK315367.1 | - | 2, 3 | 235 | |||
AY298903.1 | - | 2, 3 | 246 | |||
AY358581.1 | - | 2, 3 | 203 | |||
BC030614.2 | - | 2, 3 | 246 | 4796951 | ||
BC048810.1 | - | 2, 3 | 204 | 5243487 | ||
BP347179.1 | - | 2, 3 | 260 | |||
CD517010.1 | - | 2, 3 | 176 |