Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ?
立即註冊
產品
Antibodies
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Cell Culture Media
Chemicals
Western Blot Products
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Greener Products
查看所有產品類別
應用
生物製程
細胞培養和轉染
食品和飲料
實驗室解決方案
製藥和生物製藥
即時定量PCR
半導體分析
臨床和診斷
數位解決方案
分子光譜、同位素與元素分析
查看所有應用與技術
服務
360° CDMO 和 CRO 服務
CDMO 服务
临床试验 CRO 服务
客製化服務
企業服務
金融與租賃服務
儀器服務
實驗室資訊學
授權與商業供應
培訓服務
查看所有服務
支援
線上訂購協助
數位解決方案
產品支持
技術相關資訊
培訓與教育
查看所有協助與支援
熱門商品
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
特價商品
聯絡我們
快速訂購
訂單狀態與貨物追蹤
文件和證書
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
訂單查詢
快速訂購
登入
登入
還沒有帳戶嗎 ?
立即註冊
帳戶
訂單
Connect: Lab, Data, Apps
客製化產品與項目
儀器管理
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other BBX SIRNA Assays ›
Gene Symbol:
BBX
Gene Name:
BBX, HMG-box containing
Gene Aliases:
ARTC1, HBP2, HSPC339, MDS001
Chromosome Location:
Chr.3: 107522936 - 107811329 on Build GRCh38
UniGene ID:
Hs.124366
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s32542
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
5,366.00
Your Price
Online offer:
5366.0
Check your price ›
新增到購物車
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: BBX
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001142568.2
NP_001136040.1
16, 17
2874
NM_001276286.1
NP_001263215.1
15, 16
1865
NM_020235.6
NP_064620.2
15, 16
2784
XM_005247642.3
XP_005247699.1
16, 17
2790
XM_005247643.3
XP_005247700.1
16, 17
2802
XM_005247644.3
XP_005247701.1
17, 18
2965
XM_011513000.1
XP_011511302.1
16, 17
3002
XM_011513001.1
XP_011511303.1
16, 17
2842
XM_011513002.2
XP_011511304.1
16, 17
2796
XM_011513004.2
XP_011511306.1
15, 16
2912
XM_011513005.2
XP_011511307.1
15, 16
2706
XM_011513007.2
XP_011511309.1
15, 16
1787
XM_011513010.2
XP_011511312.1
14, 15
1697
XM_017006881.1
XP_016862370.1
18, 19
3015
XM_017006882.1
XP_016862371.1
16, 17
2875
XM_017006883.1
XP_016862372.1
14, 15
1775
GenBank mRNA
AF276948.1
-
13, 14
2572
AF454941.1
-
15, 16
2777
AF454942.1
-
13, 14
2657
AK025944.1
-
5, 6
1035
AK026082.1
-
6, 7
1206
AK130627.1
-
6, 7
860
AK291757.1
-
16, 17
2820
AK314434.1
-
16, 17
2825
AL136769.1
-
13, 14
2657
AL834307.1
-
5, 6
1094
BC012837.1
-
6, 7
1516
BC131718.1
-
15, 16
1758
40123389
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_027444, AK211657, AF454943, AF454944, AK029747, and others.
Panther Classification:
Molecular Function -
DNA-binding transcription factor
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
regulation of transcription, DNA-templated
bone development
DNA binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-55658555d6-pnd5g:80/100.66.79.83:80.
git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.41.0-Offline